Assay Details
Gene Symbol: Tbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108322.1 | NP_001101792.1 | 4 - 5 | 615 | 116 | |
XM_006248619.2 | XP_006248681.1 | 3 - 4 | 1397 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
