Assay Details
Gene Symbol: Proc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012803.1 | NP_036935.1 | 2 - 3 | 120 | 118 | |
XM_006254526.2 | XP_006254588.1 | 2 - 3 | 192 | 118 | ||
XM_006254527.2 | XP_006254589.1 | 2 - 3 | 199 | 118 | ||
XM_008772018.1 | XP_008770240.1 | 2 - 3 | 262 | 118 | ||
GenBank mRNA | BC078879.1 | - | 1 - 2 | 97 | 7122006 | 118 |
X64336.1 | - | 2 - 3 | 120 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
