Assay Details
Gene Symbol: Nrep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254595.1 | XP_006254657.1 | 1 - 2 | 101 | 81 | |
GenBank mRNA | FQ214488.1 | - | 1 - 2 | 162 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006254595.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
