Assay Details
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012576.2 | NP_036708.2 | 4 - 5 | 1812 | 65 | |
XM_008772065.1 | XP_008770287.1 | 4 - 5 | 1813 | 65 | ||
XM_008772066.1 | XP_008770288.1 | 5 - 6 | 1863 | 65 | ||
GenBank mRNA | AF455050.1 | - | 4 - 5 | 1803 | 65 | |
AY066016.2 | - | 4 - 5 | 1807 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
