Assay Details
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012576.2 | NP_036708.2 | 6 - 7 | 2078 | 117 | |
XM_008772065.1 | XP_008770287.1 | 6 - 7 | 2079 | 117 | ||
XM_008772066.1 | XP_008770288.1 | 7 - 8 | 2129 | 117 | ||
GenBank mRNA | AB115420.1 | - | 6 - 7 | 2065 | 117 | |
AF455050.1 | - | 6 - 7 | 2069 | 117 | ||
AY066016.2 | - | 6 - 7 | 2073 | 117 | ||
CB699574.1 | - | 2 - 3 | 280 | 117 | ||
M14053.1 | - | 6 - 7 | 2136 | 117 | ||
Y12264.1 | - | 6 - 7 | 2090 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
