Assay Details
Gene Symbol: Camk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012727.3 | NP_036859.2 | 2 - 3 | 170 | 66 | |
GenBank mRNA | BC128706.1 | - | 2 - 3 | 170 | 66 | |
M63334.1 | - | 64 | 66 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC128706.1, M63334.1, NM_012727.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
