Assay Details
Gene Symbol: Camk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012727.3 | NP_036859.2 | 8 - 9 | 770 | 70 | |
XM_006254523.2 | XP_006254585.1 | 7 - 8 | 712 | 70 | ||
GenBank mRNA | BC128706.1 | - | 8 - 9 | 770 | 70 | |
M63334.1 | - | 664 | 70 | |||
M64757.1 | - | 6 - 7 | 760 | 70 | ||
S65840.1 | - | 7 - 8 | 695 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
