Assay Details
Gene Symbol: Ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013059.1 | NP_001013077.1 | 1 - 2 | 202 | 98 | |
XM_006254633.2 | XP_006254695.1 | 1 - 2 | 270 | 98 | ||
GenBank mRNA | BC085887.1 | - | 1 - 2 | 202 | 7192378 | 98 |
FQ229568.1 | - | 1 - 2 | 214 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC085887.1, NM_001013059.1, XM_006254633.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
