Assay Details
Gene Symbol: Ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013059.1 | NP_001013077.1 | 4 - 5 | 507 | 128 | |
XM_006254633.2 | XP_006254695.1 | 4 - 5 | 575 | 128 | ||
GenBank mRNA | BC085887.1 | - | 4 - 5 | 507 | 7192378 | 128 |
FQ229568.1 | - | 4 - 5 | 519 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
