Assay Details
Gene Symbol: Ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013059.1 | NP_001013077.1 | 7 - 8 | 808 | 70 | |
GenBank mRNA | BC085887.1 | - | 7 - 8 | 808 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of myeloid leukocyte differentiation
negative regulation of type 2 immune response
cellular iron ion homeostasis
signal transduction
negative regulation of gene expression
positive regulation of protein ubiquitination
negative regulation of transporter activity
negative regulation of interleukin-4 production
negative regulation of T cell proliferation
positive regulation of I-kappaB kinase/NF-kappaB signaling
ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
regulation of lymphocyte differentiation
positive regulation of protein catabolic process
negative regulation of isotype switching to IgE isotypes
regulation of isotype switching to IgG isotypes
negative regulation of inflammatory response
negative regulation of protein transport
protein binding
WW domain binding
regulation of myeloid leukocyte differentiation
negative regulation of type 2 immune response
cellular iron ion homeostasis
signal transduction
negative regulation of gene expression
positive regulation of protein ubiquitination
negative regulation of transporter activity
negative regulation of interleukin-4 production
negative regulation of T cell proliferation
positive regulation of I-kappaB kinase/NF-kappaB signaling
ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
regulation of lymphocyte differentiation
positive regulation of protein catabolic process
negative regulation of isotype switching to IgE isotypes
regulation of isotype switching to IgG isotypes
negative regulation of inflammatory response
negative regulation of protein transport
protein binding
WW domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P166216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51157 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P166216 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P166216 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P166216 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P166216 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88128 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59573 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12237 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37289 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10801296 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801291 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801294 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801297 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801295 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801292 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801293 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10801290 | - | - |
Rat Genome 230 2.0 Array | 1398893_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398893_at | - | - |
Affymetrix HT Rat Focus Array | 1398893_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398893_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800002881.rn.1 | TC1800000374.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800002884.rn.1 | TC1800000374.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800002882.rn.1 | TC1800000374.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800002883.rn.1 | TC1800000374.rn.1 | RTA TC |
Rat Expression Set 230 | 1398893_PM_at | - | - |
Rat Expression Set 230 | 1398893_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801290 | - | - |
Rat Genome U34 Set | rc_AI008958_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20802286 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650482 | - | - |
