Assay Details
Gene Symbol: Slc39a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024745.1 | NP_001019916.1 | 1 - 2 | 159 | 163 | |
GenBank mRNA | BC097493.1 | - | 1 - 2 | 159 | 7461645 | 163 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC097493.1, NM_001024745.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
