Assay Details
Gene Symbol: Ammecr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107399.1 | NP_001100869.1 | 2 - 3 | 231 | 82 | |
XM_006254590.2 | XP_006254652.1 | 2 - 3 | 1045 | 82 | ||
XM_006254591.2 | XP_006254653.1 | 2 - 3 | 1082 | 82 | ||
XM_006254593.2 | XP_006254655.1 | 2 - 3 | 482 | 82 | ||
XM_008772042.1 | XP_008770264.1 | 2 - 3 | 1082 | 82 | ||
GenBank mRNA | BC168199.1 | - | 2 - 3 | 616 | 7113894 | 82 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006254593.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
