Assay Details
Gene Symbol: Ammecr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107399.1 | NP_001100869.1 | 5 - 6 | 902 | 120 | |
XM_006254590.2 | XP_006254652.1 | 5 - 6 | 1716 | 120 | ||
XM_006254591.2 | XP_006254653.1 | 5 - 6 | 1753 | 120 | ||
XM_006254593.2 | XP_006254655.1 | 5 - 6 | 1153 | 120 | ||
XM_008772042.1 | XP_008770264.1 | 5 - 6 | 1753 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
