Assay Details
Gene Symbol: Ammecr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107399.1 | NP_001100869.1 | 1 - 2 | 114 | 112 | |
XM_006254591.2 | XP_006254653.1 | 1 - 2 | 965 | 112 | ||
XM_008772042.1 | XP_008770264.1 | 1 - 2 | 965 | 112 | ||
GenBank mRNA | BC168199.1 | - | 1 - 2 | 499 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
