Assay Details
Gene Symbol: Sil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199376.1 | NP_955408.1 | 7 - 8 | 864 | 82 | |
XM_006254542.2 | XP_006254604.1 | 7 - 8 | 898 | 82 | ||
XM_008772024.1 | XP_008770246.1 | 7 - 8 | 1186 | 82 | ||
XM_008772025.1 | XP_008770247.1 | 7 - 8 | 950 | 82 | ||
GenBank mRNA | BC062050.1 | - | 7 - 8 | 864 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
