Assay Details
Gene Symbol: Stard4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106159.2 | NP_001099629.1 | 2 - 3 | 285 | 130 | |
GenBank mRNA | CB547156.1 | - | 2 - 3 | 232 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
