Assay Details
Gene Symbol: Ik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005537.1 | NP_001005537.1 | 19 - 20 | 1764 | 84 | |
GenBank mRNA | BC081870.1 | - | 19 - 20 | 1764 | 84 | |
FQ212525.1 | - | 19 - 20 | 1716 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
