Assay Details
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022209.3 | NP_071545.2 | 2 - 3 | 368 | 109 | |
NR_073588.1 | 5 - 6 | 577 | 109 | |||
GenBank mRNA | AY251277.1 | - | 2 - 3 | 320 | 109 | |
BC078834.1 | - | 5 - 6 | 577 | 7114294 | 109 | |
D14421.1 | - | 2 - 3 | 143 | 109 | ||
D38260.1 | - | 2 - 3 | 163 | 109 | ||
FM080850.1 | - | 2 - 3 | 368 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
