Assay Details
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017315.2 | NP_059011.1 | 12 - 13 | 1479 | 81 | |
XM_006254602.2 | XP_006254664.1 | 14 - 15 | 1860 | 81 | ||
GenBank mRNA | AF080452.1 | - | 12 - 13 | 1479 | 81 | |
BC078851.1 | - | 12 - 13 | 1479 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
