Assay Details
Gene Symbol: Lims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012163.1 | NP_001012163.1 | 2 - 3 | 360 | 73 | |
XM_006254596.2 | XP_006254658.1 | 2 - 3 | 497 | 73 | ||
XM_006254597.2 | XP_006254659.1 | 2 - 3 | 535 | 73 | ||
XM_008772043.1 | XP_008770265.1 | 3 - 4 | 522 | 73 | ||
GenBank mRNA | BC087108.1 | - | 2 - 3 | 360 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
