Assay Details
Gene Symbol: Lims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012163.1 | NP_001012163.1 | 3 - 4 | 417 | 52 | |
XM_006254596.2 | XP_006254658.1 | 3 - 4 | 554 | 52 | ||
XM_006254597.2 | XP_006254659.1 | 3 - 4 | 592 | 52 | ||
XM_008772043.1 | XP_008770265.1 | 4 - 5 | 579 | 52 | ||
GenBank mRNA | BC087108.1 | - | 3 - 4 | 417 | 52 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
