Assay Details
Gene Symbol: Chaf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024741.1 | NP_001019912.1 | 4 - 5 | 495 | 78 | |
XM_006248044.2 | XP_006248106.1 | 4 - 5 | 531 | 78 | ||
GenBank mRNA | BC097460.1 | - | 4 - 5 | 495 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
