Assay Details
Gene Symbol: Chaf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024741.1 | NP_001019912.1 | 8 - 9 | 872 | 107 | |
XM_006248044.2 | XP_006248106.1 | 8 - 9 | 908 | 107 | ||
GenBank mRNA | BC097460.1 | - | 8 - 9 | 872 | 7457454 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
