Assay Details
Gene Symbol: Psmg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105891.1 | NP_001099361.1 | 5 - 6 | 956 | 113 | |
XM_006248154.2 | XP_006248216.1 | 6 - 7 | 841 | 113 | ||
XM_008768571.1 | XP_008766793.1 | 6 - 7 | 1113 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
