Assay Details
Gene Symbol: Psmg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105891.1 | NP_001099361.1 | 3 - 4 | 625 | 76 | |
XM_006248154.2 | XP_006248216.1 | 4 - 5 | 510 | 76 | ||
XM_008768571.1 | XP_008766793.1 | 4 - 5 | 782 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
