Assay Details
Gene Symbol: Igsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248155.2 | XP_006248217.1 | 1 - 2 | 213 | 72 | |
XM_006248156.2 | XP_006248218.1 | 1 - 2 | 212 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006248155.2, XM_006248156.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
