Assay Details
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106791.2 | NP_001100261.2 | 11 - 12 | 1525 | 126 | |
XM_006242193.2 | XP_006242255.1 | 11 - 12 | 1593 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
protein phosphorylation
JNK cascade
cytokine-mediated signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of smooth muscle cell proliferation
magnesium ion binding
molecular_function
protein kinase activity
protein serine/threonine kinase activity
interleukin-1 receptor binding
ATP binding
cytokine production
protein phosphorylation
JNK cascade
cytokine-mediated signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of smooth muscle cell proliferation
magnesium ion binding
molecular_function
protein kinase activity
protein serine/threonine kinase activity
interleukin-1 receptor binding
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P220575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84031 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P220575 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P220575 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P220575 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P220575 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67384 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10898870 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898871 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898867 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898868 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898873 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898876 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898869 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898874 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898864 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898878 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898872 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898875 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898877 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10898862 | - | - |
Rat Genome 230 2.0 Array | 1375624_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375624_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700013204.rn.1 | TC0700001710.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700013202.rn.1 | TC0700001710.rn.1 | RTA TC |
Rat Expression Set 230 | 1375624_PM_at | - | - |
Rat Expression Set 230 | 1375624_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898862 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22275837 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360142 | - | - |
