Assay Details
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106791.2 | NP_001100261.2 | 10 - 11 | 1358 | 79 | |
XM_006242193.2 | XP_006242255.1 | 10 - 11 | 1426 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
