Assay Details
Gene Symbol: Arsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034933.2 | NP_001030105.2 | 3 - 4 | 677 | 68 | |
GenBank mRNA | BC105852.1 | - | 3 - 4 | 1058 | 7452956 | 68 |
FQ212178.1 | - | 1639 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
