Assay Details
Gene Symbol: Creld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037208.1 | NP_001032285.1 | 5 - 6 | 632 | 61 | |
GenBank mRNA | BC098722.1 | - | 5 - 6 | 632 | 61 | |
FQ229732.1 | - | 1 - 2 | 200 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
