Assay Details
Gene Symbol: Creld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037208.1 | NP_001032285.1 | 7 - 8 | 801 | 81 | |
XM_006242216.2 | XP_006242278.1 | 6 - 7 | 656 | 81 | ||
GenBank mRNA | BC098722.1 | - | 7 - 8 | 801 | 7371287 | 81 |
FQ229732.1 | - | 3 - 4 | 369 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
