Assay Details
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108105.1 | NP_001101575.1 | 10 - 11 | 1407 | 73 | |
XM_006242201.2 | XP_006242263.1 | 11 - 12 | 1269 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
