Assay Details
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108105.1 | NP_001101575.1 | 1 - 2 | 492 | 132 | |
XM_006242201.2 | XP_006242263.1 | 2 - 3 | 354 | 132 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108105.1, XM_006242201.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
