Assay Details
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108105.1 | NP_001101575.1 | 5 - 6 | 935 | 82 | |
XM_006242201.2 | XP_006242263.1 | 6 - 7 | 797 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
