Assay Details
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242180.2 | XP_006242242.1 | 1 - 2 | 478 | 63 | |
GenBank mRNA | BC085761.1 | - | 1 - 2 | 34 | 7193798 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC085761.1, XM_006242180.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
