Assay Details
Gene Symbol: Lmf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079939.1 | NP_001073408.1 | 13 - 14 | 1955 | 107 | |
GenBank mRNA | BC088333.1 | - | 3 - 4 | 368 | 107 | |
BC129102.1 | - | 13 - 14 | 1955 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
