Assay Details
Gene Symbol: Hdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053447.1 | NP_445899.1 | 3 - 4 | 490 | 140 | |
GenBank mRNA | BC161939.1 | - | 3 - 4 | 465 | 140 | |
DY318702.1 | - | 3 - 4 | 490 | 140 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
