Assay Details
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108536.1 | NP_001102006.1 | 2 - 3 | 352 | 106 | |
XM_006256528.2 | XP_006256590.1 | 3 - 4 | 270 | 106 | ||
XM_006256529.1 | XP_006256591.1 | 2 - 3 | 140 | 106 | ||
GenBank mRNA | BC158720.1 | - | 3 - 4 | 254 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108536.1, XM_006256529.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
