Assay Details
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108536.1 | NP_001102006.1 | 8 - 9 | 1160 | 77 | |
XM_006256528.2 | XP_006256590.1 | 9 - 10 | 1078 | 77 | ||
XM_006256529.1 | XP_006256591.1 | 8 - 9 | 948 | 77 | ||
GenBank mRNA | BC158720.1 | - | 9 - 10 | 1027 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
