Assay Details
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138831.1 | NP_620186.1 | 2 - 3 | 678 | 82 | |
XM_008772974.1 | XP_008771196.1 | 2 - 3 | 717 | 82 | ||
XM_008772975.1 | XP_008771197.1 | 2 - 3 | 716 | 82 | ||
GenBank mRNA | AB047324.1 | - | 2 - 3 | 678 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
