Assay Details
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138831.1 | NP_620186.1 | 3 - 4 | 1132 | 96 | |
XM_008772974.1 | XP_008771196.1 | 3 - 4 | 1171 | 96 | ||
GenBank mRNA | AB047324.1 | - | 3 - 4 | 1132 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
