Assay Details
Gene Symbol: Nt5dc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106393.1 | NP_001099863.1 | 11 - 12 | 1358 | 110 | |
XM_008772980.1 | XP_008771202.1 | 11 - 12 | 1374 | 110 | ||
XM_008772981.1 | XP_008771203.1 | 11 - 12 | 1371 | 110 | ||
GenBank mRNA | BC162021.1 | - | 11 - 12 | 1322 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P385105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17507 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P440988 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P440988 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P440988 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P147426 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P440988 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94336 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94032 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37953 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66195 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23562 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10833583 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833572 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833576 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833580 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833573 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833584 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833574 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833577 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833581 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833578 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833582 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833579 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833575 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10833571 | - | - |
Rat Genome 230 2.0 Array | 1392144_at | - | - |
Rat Genome 230 2.0 Array | 1392144_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392144_at | - | - |
Affymetrix HT Rat Focus Array | 1392144_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000013423.rn.1 | TC2000001639.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR2000013424.rn.1 | TC2000001639.rn.1 | RTA TC |
Rat Expression Set 230 | 1392144_PM_at | - | - |
Rat Expression Set 230 | 1392144_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833571 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21440717 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364225 | - | - |
