Assay Details
Gene Symbol: Nt5dc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106393.1 | NP_001099863.1 | 6 - 7 | 646 | 118 | |
XM_008772980.1 | XP_008771202.1 | 6 - 7 | 662 | 118 | ||
XM_008772981.1 | XP_008771203.1 | 6 - 7 | 659 | 118 | ||
GenBank mRNA | BC162021.1 | - | 6 - 7 | 610 | 9098589 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106393.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
