Assay Details
Gene Symbol: Nt5dc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106393.1 | NP_001099863.1 | 8 - 9 | 924 | 72 | |
XM_008772980.1 | XP_008771202.1 | 8 - 9 | 940 | 72 | ||
XM_008772981.1 | XP_008771203.1 | 8 - 9 | 937 | 72 | ||
GenBank mRNA | BC162021.1 | - | 8 - 9 | 888 | 9098589 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
