Assay Details
Gene Symbol: Nt5dc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106393.1 | NP_001099863.1 | 9 - 10 | 1040 | 66 | |
XM_008772980.1 | XP_008771202.1 | 9 - 10 | 1056 | 66 | ||
XM_008772981.1 | XP_008771203.1 | 9 - 10 | 1053 | 66 | ||
GenBank mRNA | BC162021.1 | - | 9 - 10 | 1004 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
