Assay Details
Gene Symbol: Mfsd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108730.1 | NP_001102200.1 | 3 - 4 | 672 | 169 | |
XM_008765107.1 | XP_008763329.1 | 3 - 4 | 740 | 169 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
