Assay Details
Gene Symbol: Gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012793.2 | NP_036925.2 | 5 - 6 | 624 | 106 | |
GenBank mRNA | FQ225896.1 | - | 5 - 6 | 624 | 106 | |
FQ227114.1 | - | 5 - 6 | 624 | 106 | ||
J03588.1 | - | 5 - 6 | 622 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
