Assay Details
Gene Symbol: Dos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108070.1 | NP_001101540.1 | 5 - 6 | 877 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108070.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
