Assay Details
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109882.1 | NP_001103352.1 | 4 - 5 | 751 | 100 | |
NM_012783.3 | NP_036915.1 | 3 - 4 | 403 | 100 | ||
GenBank mRNA | AY120888.1 | - | 4 - 5 | 670 | 100 | |
BC059145.1 | - | 3 - 4 | 327 | 6889785 | 100 | |
FQ212571.1 | - | 3 - 4 | 339 | 100 | ||
FQ212582.1 | - | 3 - 4 | 347 | 100 | ||
FQ213065.1 | - | 3 - 4 | 349 | 100 | ||
FQ213626.1 | - | 3 - 4 | 339 | 100 | ||
FQ214193.1 | - | 3 - 4 | 339 | 100 | ||
FQ214232.1 | - | 3 - 4 | 344 | 100 | ||
FQ218399.1 | - | 3 - 4 | 339 | 100 | ||
FQ219910.1 | - | 3 - 4 | 323 | 100 | ||
FQ220920.1 | - | 3 - 4 | 347 | 100 | ||
FQ224837.1 | - | 3 - 4 | 348 | 100 | ||
FQ229144.1 | - | 3 - 4 | 349 | 100 | ||
FQ229755.1 | - | 3 - 4 | 336 | 100 | ||
FQ230014.1 | - | 3 - 4 | 349 | 100 | ||
FQ231850.1 | - | 3 - 4 | 349 | 100 | ||
FQ232262.1 | - | 3 - 4 | 339 | 100 | ||
FQ234809.1 | - | 3 - 4 | 347 | 100 | ||
X54640.1 | - | 530 | 100 | |||
X67215.1 | - | 3 - 4 | 324 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
