Assay Details
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109882.1 | NP_001103352.1 | 6 - 7 | 1172 | 119 | |
NM_012783.3 | NP_036915.1 | 5 - 6 | 824 | 119 | ||
GenBank mRNA | AY120888.1 | - | 6 - 7 | 1091 | 119 | |
BC059145.1 | - | 5 - 6 | 748 | 119 | ||
BQ199464.1 | - | 2 - 3 | 119 | |||
FQ212571.1 | - | 5 - 6 | 760 | 119 | ||
FQ213065.1 | - | 5 - 6 | 770 | 119 | ||
FQ214232.1 | - | 5 - 6 | 765 | 119 | ||
FQ218399.1 | - | 5 - 6 | 759 | 119 | ||
FQ220920.1 | - | 5 - 6 | 768 | 119 | ||
FQ224837.1 | - | 5 - 6 | 769 | 119 | ||
FQ229144.1 | - | 5 - 6 | 770 | 119 | ||
FQ229755.1 | - | 5 - 6 | 757 | 119 | ||
FQ231850.1 | - | 5 - 6 | 770 | 119 | ||
FQ232262.1 | - | 5 - 6 | 760 | 119 | ||
FQ234420.1 | - | 5 - 6 | 770 | 119 | ||
FQ234809.1 | - | 5 - 6 | 119 | |||
X54640.1 | - | 951 | 119 | |||
X67215.1 | - | 5 - 6 | 745 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
